Glass Roller Culture Vessels, Tapered Neck, with Phenolic Caps, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Vessels are made to exacting tolerances to provide higher cell yields.
- Vessels and deep-skirted 38-415 screw caps are autoclavable
- Free of glass imperfections
- Neck area is tapered to facilitate cleaning
- Caps have white styrene-butadiene rubber liner
Glass is specially selected for optical clarity and interior surface smoothness.
Certifications: Wheaton-33® low extractable borosilicate glass meets ASTM Type I Class A and USP Type I standards.
Customers Who Bought This Also Bought