Amplitude™ Theta™ Wipes, Contec®
Supplier: Contec
Certificates
About this item
Amplitude™ Theta™ Wipes are made from a lightweight hydroentangled cellulose/polyester, which creates an economical absorbent wipe ideal for general cleaning applications.
- Recommended for ISO Class 6 to 8 environments
- Economical and cost-effective nonwoven wipe
- Lightweight sorbent wipe
- Double packaged to preserve product integrity
These wipes offer excellent durability and sorbency. They are designed for nonabrasive applications, such as general-purpose wiping and polishing of surfaces.
Related Information
Certificates
Search by Lot Number89030-190_172695 89030-190_173276 89030-190_178068 89030-190_179390 89030-192_171771
Conformity declaration
Customers Who Bought This Also Bought