Terminal® Broken Glass Benchtop Keeper™ Container, Whitney Products
Catalog #: 11214-713
Supplier: Whitney Products
Specifications
- Capacity:3 L (101.4 oz.)
- Dimensions:15.2L×15.2W×25.4H cm (6×6×10")
- Cat. no.:11214-713
- Supplier No.:BH2004
Specifications
About this item
For the safe collection of non-biohazardous broken glass
- Contents are clearly identified
- Plastic-coated lining for leak resistance
- Self-locking container is autoclavable
For the safe collection of non-biohazardous broken glass
Helps protect against accidental injuries.
Customers Who Bought This Also Bought