Amplitude™ Zeta™ Wipes, Contec®
Supplier: CONTEC, INC. MX
Certificates
About this item
Amplitude™ Zeta™ wipes are a 5-ply, thermally bonded wipe which is comprised of a layer of cellulose sealed between two layers of meltblown polypropylene. This creates a material which is very absorbent and has a high level of cleanliness.
- Recommended for ISO Class 6 to 8 environments
- 5-ply quilted pattern allows for segregation by appearance
- High level of cleanliness for a general-purpose wipe
- Ideal for solvent pick up and wiping to dry
- Double packaged to preserve product integrity
- Good chemical compatibility, including acids
- Can be used as tray liners and spill mats
The wipes have good chemical compatibility and are resistant to most disinfectants and solvents, including acids. Absorbent enough for solvent and acid spills, they are also ideal for wiping to dry applications.
Amplitude™ Zeta™ wipes make useful tray liners and are suitable for general and lab area applications.
Related Information
Certificates
Search by Lot Number89030-194_171773 89030-194_172371 89030-194_172699 89030-194_178063 89030-194_179361
Conformity declaration
More
Customers Who Bought This Also Bought