Specifications
- Capacity:60.6 L (16 gal.)
- Dimensions:0.5W x 30.5D x 68.6H cm (12 x 12 x 27")
- Cat. no.:12000-800
- Supplier No.:BH2009
Specifications
About this item
Floor model ideal for the disposal of non-sharp biohazardous lab waste, including petri dishes, tubes, plates, pipets and pipet tips, specimen cups, and tubing sets
Unique Reclose-A/Remove-A-Lid™ feature lets users reclose the container to reduce odors and mess or remove the lid entirely. The wide-mouth opening allows disposal of large items, or entire racks of tubes or pipet tips at once. Containers store flat, conserving storage space, and assemble easily in seconds. Container is marked with the biohazard symbol and includes warnings in English and Spanish.
Floor model ideal for the disposal of non-sharp biohazardous lab waste, including petri dishes, tubes, plates, pipets and pipet tips, specimen cups, and tubing sets
Unique Reclose-A/Remove-A-Lid™ feature lets users reclose the container to reduce odors and mess or remove the lid entirely. The wide-mouth opening allows disposal of large items, or entire racks of tubes or pipet tips at once. Containers store flat, conserving storage space, and assemble easily in seconds. Container is marked with the biohazard symbol and includes warnings in English and Spanish.
Sturdy corrugated paperboard construction with heavy-duty 2mil red plastic liner for leak resistance.