Terminal® Safe Keeper™ Waste Container, Whitney Products
Catalog #: 11214-717
Supplier: Whitney Products
Specifications
- Capacity:1.1 L (37.2 oz.)
- Dimensions:14.6L×5.1W×27.9H cm (5³/₄×2×11")
- Cat. no.:11214-717
- Supplier No.:BH2005
Specifications
About this item
For the safe collection of biohazardous items such as capillary tubes, pipet tips, swabs, and more.
- Equipped with reinforced hole for wall-hanging
- Completely closed bottom to avoid leakage
- Self-adhesive hook is included in each case
- Constructed of sturdy, plastic-lined paperboard for leak resistance
- Autoclavable
For the safe collection of biohazardous items such as capillary tubes, pipet tips, swabs, and more.
Customers Who Bought This Also Bought