Terminal® Pipet Keeper™ Disposal Container, Whitney Products
Catalog #: 11214-715
Supplier: Whitney Products
Specifications
- Dimensions:39.4L×7.6W×7.6H cm (15¹/₂×3×3")
- Cat. no.:11214-715
- Supplier No.:BH2006
Specifications
About this item
For the collection of biohazard waste at the point of generation.
For the collection of biohazard waste at the point of generation. Holds pipets, swabs, and other objects up to 38.1cm (15") long. Constructed of sturdy, plastic-lined paperboard for leak resistance. Autoclavable container has a reclosable flap. Low profile prevents tipping.
- Constructed of sturdy, plastic-lined paperboard for leak resistance
- Autoclavable container has a reclosable flap
- Low profile prevents tipping
Holds pipets, swabs, and other objects up to 38.1cm (15") long.
Customers Who Bought This Also Bought