About this item
Low-linting wipers for ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom assembly operations, pharmaceutical operations, and other clean environments.
Low-linting wipers for ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom assembly operations, pharmaceutical operations, and other clean environments. Made of a unique, 100% polypropylene, nonwoven fabric that is low in particulate matter and low in extractable levels. Soft, absorbent, durable wipers contain no binders or reinforcing fibers. They eliminate surface fibers, trap particles, and will not scratch sensitive surfaces. Each wiper can absorb over four times its weight in liquids. Acid-, base-, and solvent-resistant; will not break down or disintegrate when exposed to chemicals. Color: white.
- 100% polypropylene, non-woven fabric is low in particulate matter and extractable levels
- Soft, absorbent, durable
- Contain no binders or reinforcing fibers
- Eliminiate surface fibers, trap particles, will not scratch surfaces
- Color: white
Each wiper can absorb over four times its weight in liquids. Acid-, base-, and solvent-resistant; will not break down or disintegrate when exposed to chemicals.
For ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom assembly operations, pharmaceutical operations, and other clean environments.
Related Information
Certificates
Search by Lot Number21908-205_CO3A 83009-302_CO31 83009-302_CO3D 83009-302_CO3E