ContactUsLinkComponent
EPA method 624 Standards Kit, Restek
EPA method 624 Standards Kit, Restek
Catalog #: RK30055
Supplier:  Restek
CAS Number:  
EPA Method 624 Standards Kit
EPA method 624 Standards Kit, Restek
Catalog #: RK30055
Supplier:  Restek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EPA 624 Kit (Mixtures No. 1, 2, 3 and Internal Standard Mixture)
  • Cat. no.:
    RK30055
  • Supplier No.:
    30055
Specifications
About this item
Calibration mixes are packaged in purge and trap methanol at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Contents of 624 Calibration Mixture No. 2: Benzene, Carbon Tetrachloride, Chlorobenzene, 2-Chloroethyl Vinyl Ether, Dibromochloromethane, 1,1-Dichloroethane, 1,1-Dichloroethene, 1,2-Dichloropropane, Methylene Chloride, Tetrachloroethylene, 1,1,2-Trichloroethane, and Trichloroethene
Internal standard mix is packaged in purge and trap methanol at a concentration of 1500 µg/mL in a 1 mL ampoule. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Customers Who Bought This Also Bought