About this item
Molded from CarmelStat™, these black ESD AkroBins® bins dissipate electrostatic charges for safe and secure storage of sensitive electronic components
Unaffected by weak acids and alkalis. Sturdy, heavy-duty, one-piece construction is water, rust, and corrosion-proof. Resistant to oil and grease.
- Unaffected by weak acids and alkalis
- Sturdy, heavy-duty, one-piece construction is water, rust, and corrosion-proof
- Resistant to oil and grease
Molded from CarmelStat™, these black ESD AkroBins® bins dissipate electrostatic charges for safe and secure storage of sensitive electronic components
Containers feature a full-width hanger lip to support heavy loads when suspended from hanging rails or louvered panels. Reinforced side ribs provide added strength to prevent side-wall spreading when bins are stacked or loaded. Large label slot on front of bin provides easy bar code scanning and content identification. Anti-slide stop prevents bins from shifting forward when stacked.