Specifications
- Description:Model A6 System
- Cat. no.:27372-131
- Supplier No.:A6
Specifications
About this item
Wide-format system offers a simple, convenient, and fast method for screening up to 500 samples simultaneously on a single agarose gel
The 23Wx25Lcm (9x913/16") gel tray is supplied with gasketed end plates for leak-free gel casting without tape. Comb slot design allows the use of up to 10 combs spaced evenly along the tray for maximum sample capacity. Samples may be loaded directly from a 96- or 384-well plate, speeding loading time and reducing the chance of sample order error. System dimensions: 37.1Lx29.7Wx10.2Hcm (145/8x1111/16x4").
System is built on a leveling platform to ensure the casting and running of flat gels to produce clear, tight banding patterns without “smiling.”