Nunc® Labtop Cooler, Thermo Scientific
Catalog #: CA13500-400
Supplier: Thermo Fisher Scientific
Specifications
- Description:Labtop Cooler
- Cat. no.:CA13500-400
- Supplier No.:355501
Specifications
About this item
Designed specifically to protect enzymes, reagents, cells, and solutions by maintaining temperatures between –20°C and –15°C for up to four hours
- Printed grid helps to inventory tubes while the keyed lid maintains proper orientation
- Locking handle for safe and easy transport
- Nonskid feet on bottom prevent cooler from sliding on the benchtop
Break-resistant polycarbonate cooler holds twenty 1.5–2.0mL microcentrifuge tubes or NUNC® CryoTube® vials. Included inserts hold 0.2–0.5mL microcentrifuge tubes.
Customers Who Bought This Also Bought