Glass Plates for Owl™ Aluminum Backed Sequencer System, Models S1S, S2S, S3S and S4S, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
Glass Plates for Owl™ Aluminum Backed Sequencer System, Models S1S, S2S, S3S and S4S, Thermo Scientific
Supplier: Thermo Fisher Scientific
About this item
Plates are available in blank, notched, or offset glass.
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$64.40 to $734.93
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Listeria Palcam Selective Enrichment Broth Base Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
$663.68 to $3,804.95