Horizon Educational DIY Fuel Cell Science Kit
Supplier: HORIZON FUEL CELL TECHNOLOGIES
New Product
Certificates
About this item
Organize a DIY race, fueled by hydrogen generated electricity!
- All-in-one electrolyzer and fuel cell
- Activity guide
- Free online curriculum resources
- DIY chassis kit attachment available for separate purchase
Students are tasked with designing and fabricating a car that is powered by H₂. They will design their vehicle, source materials for the fuel cell system, build a prototype, and test it in a race.
Educational concepts covered include climate change, renewable energy, human impact, electrolysis, energy, hydrogen generation, reaction rates, reaction yield, reactions, redox reactions, classical mechanics, efficiency, electric circuits, energy, Ohm's law, power (electric).
Delivery: Includes reversible fuel cell, silicone tubing, syringes, LEDs, wires, pins, battery pack, motor, gear box, reief valves, and clamps.
Customers Who Bought This Also Bought