Specifications
- Description:50 ml bottle
- Supplier No.:27131
Specifications
About this item
Designed for the permanent locking and sealing of fasteners such as large bolts and studs (M25 and larger) up to 25mm (1") diameter.
- Acrylic threadlocker
- High-strength
- Low-viscosity
The adhesive cures when confined in the absence of air between close fitting metal surfaces and prevents loosening and leakage from shock and vibration. Low viscosity reduces the migration of liquid adhesive after application.