Specifications
- Description:Super Bonder® 409™ industrial grade instant adhesive gel
- Cat. no.:83007-506
- Supplier No.:40945
Specifications
About this item
Clear adhesive gel for general-purpose use on metal, plastic, and rubber.
- Clear instant adhesive
- Thixotropic gel formulation
- Prevents adhesive runoff or drip
The thixotropic gel formulation fills large gaps and is useful for overhead or hard-to-reach applications where adhesive runoff or drip must be avoided.