Laboratory Fume Hoods, A User’s Manual, Houghton Mifflin
Catalog #: 56615-572
Supplier: Houghton Mifflin
Specifications
- Description:Laboratory Fume Hoods, A User’s Manual by G. Thomas Saunders
- Cat. no.:56615-572
- Supplier No.:0471-56935-6
Specifications
About this item
Written for industrial hygienists, safety engineers, toxicologists, industrial psychologists, and public health professionals.
- Thorough reference on adequate fume hood design and use dissects the device down to its bare essentials
- Examines how and why a fume hood works
- Helps the user test, locate, ventilate and maintain hoods (both old and new) which are on site or field-generated
144 pages, released 1993
Customers Who Bought This Also Bought