Storage and Assay Microplates
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
New Product
Certificates
About this item
For biological assays, low storage temperatures, and filtrate collection applications.
- Compatible with all major automation platforms and benchtop devices
- No risk feasibility testing – test a sample in your application
- Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
- Molded with chemical resistant polymers, for maintained performance with prolonged exposure to aggressive solutions and solvents
- Sealable with heat and adhesive seals, or cap mats, for superior performance
- Low temperature tolerant for refrigerator or freezer storage
- Autoclavable, can be sterilized at 121 °C, 15 psi for 20 minutes
- Available in a large range of well volumes, from 58 µl to 10 ml for application scale-up or scale-down
- Complementary fit to Agilent filter microplates for centrifugation stacking
- Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
Agilent supplies collection and storage assay microplates for a wide range of reaction chemistries, biological assays, low storage temperatures, and filtrate collection applications. These microplates are molded with medical-grade, chemical-resistant polymers and are available in 24, 48, 96, or 384-well configurations of variable volumes. Agilent also supplies mat seals and lids for these microplates.
Customers Who Bought This Also Bought