KIMBLE® KIMAX® Gooch Crucibles, Low Form, with KIMFLOW® Fritted Disc, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Especially suitable for precipitates to be dried to constant weight at 110°C
Note: Exposure to sudden temperature changes should be avoided.
For higher temperatures, an electric furnace is advised and crucibles should not be removed until temperature has dropped to 250°C.