Tissue Cassettes, Round Hole, Without Lid
Catalog #: 76809-040
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
Specifications
- Color:White
- Description:Round hole cassettes, without lid
- Packaging:Bulk
Specifications
About this item
Round hole processing cassettes ensure precise tissue processing with minimized deformation.
- For use with reusable metal stainless steel lids (sold separately)
- 45° angled writing surface for convenient labeling
- For use with laser printers
Crafted for optimal fluid exchange during processing, these round hole cassettes maximize efficiency in tissue preparation.
Packaging: Bulk packaging: 500 per bag, 10 bags per case.
Customers Who Bought This Also Bought