Biopsy Cassettes, Sleeved, Two-Piece
Catalog #: 76809-034
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
New Product
Specifications
- Color:White
- Description:Two-piece biopsy cassettes
- Packaging:Sleeved
Specifications
About this item
POM biopsy cassettes ensure precise tissue embedding and sectioning in histology labs.
- POM material minimizes wax block deformation
- Even and uniform printing surfaces
- Special hinge allows easy cover removal
- 45° angled writing surface for convenient labeling
- For use with laser printers
Biopsy cassettes crafted from POM material resist deformation caused by temperature variations during tissue processing. The uniform printing surfaces ensure precise labeling with text and data matrix codes. Users can effortlessly open and close the hinged covers for efficient workflow. The detachable cover can be easily removed and re-attached as many times as desired.
Packaging: Sleeved packaging: 90 per sleeve, 20 sleeves per case.
Customers Who Bought This Also Bought