Specifications
- Color:Silver
- Description:Zinc plated steel binClips for DuraBoard® or ¹/₈" and ¹/₄" pegboard, 5 per pack
Specifications
About this item
The patented DuraHook® is the world’s first double locking, multi-position pegboard hook and provides a secure, innovative hanging storage solution.
- Plastic hanging bins mount on top of the BinClip which secures the bin to the DuraBoard
- Use on any ¹/₈" or ¹/₄" pegboard (excluding steel)
- Bins are easily removed from the BinClip, taken to work areas and then returned to the BinClip position on the DuraBoard when the job is finished
- Use multiple steel BinClips for larger bins and heavier items
Triton BinClip is a unique design that makes it easy to mount plastic hanging bins on all pegboard and DuraBoard sizes. They provide additional storage opportunities on a pegboard for those hard to store parts and tool accessories. LocBin's or any plastic hanging bin can be mounted on the BinClip individually anywhere on the pegboard to securely hold loose parts, tool accessories or heavy, odd-shaped or precision tools along with hardware items. Pegboard BinClips provide a visible and interchangeable bin system for any size pegboard and offer a unique storage opportunity to keep accessory items and small parts neat and organized near the tools that are being stored on a pegboard.