DuraBoard® Pegboard with 36-Piece DuraHook® Assortment
Supplier: Triton Products
New Product
Certificates
About this item
DuraBoard® is the most durable, weather and impact resistant pegboard available. It is eight times stronger than conventional pegboard.
- Hole diameter is ¹/₄" with 1" O.C spacing
- DuraBoard will never fade, dry out, crack or peel over time so it maintains holding capability over many years of use
- Offers a professional looking pegboard system that can be cleaned without harming the board
- Board surface is ideal for creating silhouettes of tools to be stored and managed allowing for easy tool identification
- DuraHooks are easily repositioned in seconds as requirements change with no damage to the board
The commercial-grade DuraBoard® ¹/₄" pegboard has superior holding strength with eight times that of traditional pegboard. It is the most durable, weather and impact resistant pegboard available. This pegboard will stand up to any harsh temperatures and environments and can be cleaned or washed when necessary. DuraBoard® heavy-duty pegboard is ideal for use in a clean room, factory floor production area, maintenance and repair area as well as sheds and garages. Use DuraHook® double locking pegboard hooks to create the most secure and reliable pegboard storage system.
Ordering information: Limited lifetime warranty.
Delivery: Includes (1) 24×48×¹/₄" DuraBoard; (6) 1" Single rod 30° bend; (1) 6" Single rod 30° bend; (6) 2¹/₂" Single rod 30° bend; (4) Pliers/Wrench holder; (5) Extended spring clip; (4) Standard spring clips; (5) Single ring tool holder; (5) Curved hooks and mounting hardware.
Customers Who Bought This Also Bought