Specifications
- Color:Silver
- Description:Pegboard hook, 2³/₄" double rod 80 degree bend, stainless steel, for ¹/₈" and ¹/₄" pegboard, 3 per pack
Specifications
About this item
The Stainless Steel DuraHook® is ideal for food, medical, healthcare, restaurant, marine and commercial applications as well as any clean room or harsh environment that must meet stringent requirements.
- Ideal for storing hammers, sledge hammers, large hand tools and power tools when used with other stainless steel DuraHooks
- Innovative double locking system locks the hook in place with a locking tab that mounts to the pegboard from the front
- Won't rust or corrode in any environment, even ones with extreme temperatures
- Use on any ¹/₈" or ¹/₄" pegboard (excluding steel)
- Hooks mount vertically and horizontally in 4 distinct positions allowing the use of multiple hooks for large, heavy and odd-shaped items
- Fixtures install from the front, so no need to remove the board for installation or repositioning of stainless steel DuraHooks
Having clean, easily accessible and versatile storage and organizational solutions is an issue that every business encounters. The food and healthcare industries must meet stringent requirements and the stainless steel pegboard storage from Triton Products provides the ideal answer. Our innovative DuraHook is now available in stainless steel. The Stainless Steel DuraHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. Imagine a clean, well-kept kitchen with every pan, ladle, knife and utensil conveniently hanging and within grasp at all times. Imagine every medical supply in a hospital being easily accessible at a moment's notice. That is the power of the stainless steel DuraHook from Triton Products.