Corning® Spinner Flasks with Angled Sidearms, Corning
Supplier: Corning
Certificates
About this item
Borosilicate glass with side arms, non sterile.
- Baffles enhance aeration and agitation of flask contents
- Impeller design ensures optimal stirring
- Complete with impeller and caps
- Side arm design permit easy access of 25 and 50 ml pipetes
Customers Who Bought This Also Bought