Pierce SuperSignal™ ELISA Femto Substrate, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
SuperSignal® ELISA Femto maximum sensitivity substrate utilizes the SuperSignal® system with an enhancer formulation for superior protein detection and low-end linearity in ELISA applications.
- Intense signal generated immediately at both room temperature and 37 °C
- Easy detection of low quantities of proteins with high signal:noise ratios and low-end linearity of dose response curves
- Femtogram-level detection of target proteins in ELISA
- High sensitivity allows for reduction in ELISA incubation steps and hence a reduction in assay time
- Storage at room temperature with a 6 hour working solution stability
- Ideal for automated high-throughput screening applications
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.