Order Entry
ContactUsLinkComponent
VWR® Large Wide Mouth Square Bottle, HDPE
VWR® Large Wide Mouth Square Bottle, HDPE
Catalog #: 414004-190
Supplier:  VWR International
VWR Plastic Bottles
VWR® Large Wide Mouth Square Bottle, HDPE
Catalog #: 414004-190
Supplier:  VWR International

Specifications

  • Capacity:
    4 L (1 gal.)
  • Dimensions:
    27.9H x 14D x 14W cm (11 x 51/2 x 51/2")
  • Cat. no.:
    414004-190

Specifications

About this item

Leakproof, space-saving 3.8 L (1 gal.) bottle is ideal for collecting, storing, and transporting dry chemicals, environmental samples, reagents, and distilled water.

  • Leakproof
  • Space-saving design
  • Wide mouth for easy access to contents

Can be used for low-temperature applications and stored in freezer to −100 °C (−148 °F). Features integrated handle for better grip, greater safety, and pouring convenience.

Ordering information: 100-415 white polypropylene cap included, assembled.
Customers Who Bought This Also Bought