KIMAX® Graduated Cylinders, Class B, To Deliver, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Single metric scale is durable white ceramic enamel
Designed from ASTM Specification E1272, Style 1, Class B requirements.
Hexagonal base provides stability. The 10mL size has funnel top and sizes 25mL and larger have SAFE-GARD® bumpers. The 100mL size may be used for some ASTM distillation tests.
Specifications
- Catalog No:
- 89001-122
- 89001-124
- 89001-118
- 89001-120
- 89001-114
- 89001-116
- 89001-110
- 89001-112
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
Specifications
Related Information
Certificates
Search by Lot Number89001-114_220185676 89001-118_220183279 89001-122_220182732 89001-124_220181650 89001-124_220183897
Conformity declaration
DWK Kimble Silicone Lid Brochure
Product Literature
DWK Life Sciences Return to Research Promo 2021
Promotion Material
Customers Who Bought This Also Bought