About this item
CD16 (Clone 3G8) is a mouse monoclonal antibody that recognizes IgG receptor CD16, also known as FC gamma receptor III. This antibody belongs to the Biotium Choice list of select antibodies that have been validated and optimized in-house for optimal performance. The antibody is available conjugated to CF® Dyes and Astral™ tandem dyes. They are supplied in PBS, 0.1% BSA, 0.05% azide.
- Cell/tissue expression: Lymphocytes
- SwissProt: P08637
- Unigene: 372679
- Positive control: Human peripheral blood mononuclear cells (PBMCs)
- Ex/Em: 496, 546, 565/576 nm
- Storage condition: Protect fluorescent conjugates from light
This antibody recognizes CD16 (Fc gamma receptor III), the low-affinity receptor for IgG with an apparent molecular weight of 50 to 80 kDa. CD16 is represented by two similar genes, CD16A (Fc gamma receptor IIIA), which exists as a hetero-oligomeric polypeptide-anchored form in macrophages and NK cells and CD16B (Fc gamma receptor IIIB), which exist as a monomeric GPI-anchored form in neutrophils. Furthermore, there are two known polymorphisms of CD16B, NA-1 and NA-2. Individuals homozygous for NA-2 show a lower phagocytic capacity compared with NA-1.
CD16 binds IgG in the form of immune complexes and shows preferential binding of IgG1 and IgG3 isotypes and minimal binding of IgG2 and IgG4. Upon IgG binding, both CD16 isoforms initiate signal transduction cascades that lead to a variety of responses including antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, degranulation and proliferation.
Shelf life: Guaranteed for at least 24 months from date of receipt when stored as recommended.
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: 3G8
Conjugation: R-PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77449-090
- 77449-092
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 3G8
- 3G8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 2214
- 2214
- Isotype:
- IgG1 kappa
- IgG1 kappa