VITLAB® Reagent Bottles, PFA, Wide Mouth, BrandTech
Supplier: Brandtech
Certificates
About this item
Leach-resistant bottles are for long-term storage of solvents and standards.
- Thread finish: S 40
- Bottles and caps are autoclavable
Bottles are made of perfluoroalkoxy copolymer (PFA), which is highly resistant to heat and chemical contamination. PFA’s smooth surface and hydrophobic, anti-adhesive properties simplify the cleaning process, making it ideal for trace analysis. Bottles maintain stability in temperatures from –200 to 260°C (–328 to 500°F). Supplied with PFA screw cap with a molded sealing ring.
Customers Who Bought This Also Bought