Leak-Resistant Bottles, Low-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Lightweight bottles are ideal for general laboratory use.
- Wide mouth facilitates filling and pouring
- Bottles are designed with a special pour lip to facilitate safe, clean dispensing
- Double-seal closure for leak resistance
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.
Customers Who Bought This Also Bought