Leak-Resistant Bottles, Polypropylene, Wide Mouth, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Designed for general laboratory use.
- Wide mouth facilitates easy filling and pouring
- Equipped with a special pour lip to facilitate safe, clean dispensing
- Double-seal closure for leak resistance
- Autoclavable
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.