Order Entry
ContactUsLinkComponent
Pierce™ Glycoprotein Staining Kit, Thermo Scientific
Pierce™ Glycoprotein Staining Kit, Thermo Scientific
Catalog #: CAPI24562
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ Glycoprotein Staining Kit, Thermo Scientific
Catalog #: CAPI24562
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Pierce™ Glycoprotein Staining Kit
  • Size:
    1 Kit
  • Cat. no.:
    CAPI24562
  • Supplier No.:
    24562

Specifications

About this item

The Pierce Glycoprotein Staining Kit is a convenient, fast, and sensitive colorimetric kit for staining glycosylated proteins in polyacrylamide gels or nitrocellulose membranes using the periodic acid-Schiff (PAS) method.

  • Specific for glycoproteins—PAS method specifically detects glycosylated proteins with sialic acid and other oxidizable carbohydrate groups
  • Protocol—the traditional three-reagent method has been streamlined to achieve optimal staining in less than two hours
  • Colorimetric—glycoproteins are detected as magenta bands against a faint pink or colorless gel background
  • Qualitative staining—detection limit depends on the extent of glycosylation and protein size and abundance; detects 0.625 ng avidin and 0.16 µg horseradish peroxidase per band
  • Controls included—complete kit includes positive and negative control proteins (HRP and STI, respectively).

The kit provides three essential reagents and a complete protocol to specifically stain glycosylated proteins (glycoproteins) that have been separated by polyacrylamide gel electrophoresis (PAGE). A gel or membrane containing separated proteins is treated with a periodate solution, which oxidizes cis-diol sugar groups in glycoproteins. The resulting aldehyde groups are detected through the formation of Schiff-base bonds with a reagent that produces magenta bands. Traditionally called the periodic acid-Schiff (PAS) reagent method, this staining technique is made easier by the convenient packaging of essential reagents in one kit with easy-to-follow instructions.

Contents
Sufficient For: 10 mini gels (SDS-PAGE)
• Oxidation Reagent, 2.5 g powder to make 250 mL
• Reduction Reagent, 1.25 g powder to make 250 mL
• Positive Control (Horseradish Peroxidase), 1 mg
• Negative Control (Soybean Trypsin Inhibitor), 1 mg
• Glycoprotein Stain, 250 mL

Ordering information: The 6xHis protein tag staining kit (PI24575) and phosphoprotein staining kit (PI24550) come with the appropriate reagent and control sets, which can be purchased separately.
Caution: Store all components at 4°C. For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought