Order Entry
ContactUsLinkComponent
SP Bel-Art Separatory Funnel and Imhoff Cone Rack, Bel-Art Products, a part of SP
SP Bel-Art Separatory Funnel and Imhoff Cone Rack, Bel-Art Products, a part of SP
Catalog #: 30534-905
Separatory Funnel Rack
SP Bel-Art Separatory Funnel and Imhoff Cone Rack, Bel-Art Products, a part of SP
Catalog #: 30534-905

Specifications

  • Dimensions:
    22L x 66W x 74H cm
  • Description:
    Imhoff Cone and Separatory Funnel Rack
  • Cat. no.:
    30534-905
  • Supplier No.:
    189670000

Specifications

About this item

This high-density polyethylene rack holds four separatory funnels on the lower shelf or four long-stem funnels on the upper shelf (maximum funnel diameter is 98 mm).

Set screws allow shelf height adjustments anywhere along the 74 cm support rods.

Delivery: Rack includes 24 insert clips that allow the rack to accommodate different sized separatory funnels and imhoff cones.
Customers Who Bought This Also Bought