SP Bel-Art HOLDFAST™ Graduated Cylinders, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
This one-piece cylinder with handle provides a secure, solid grip for safer pouring.
- Polypropylene construction
- Secure, solid grip with handle
- Easy-to-read graduations
- Chemically resistant
- Autoclavable
These cylinders have an easy-to-read double scale with raised graduations, and a hexagonal base to prevent tipping and rolling. Made of polypropylene, it will not create a meniscus. Handle opening is 8.9 x 2.9 cm (3 x 1⅛"). Chemically resistant and autoclavable at 121ºC (250ºF).
Specifications
- Catalog No:
- 24780-369
- 24780-371
- Form:
- Tall
- Tall
Specifications
Related Information
Certificates
Search by Lot NumberBel-Art Cleaning and General Supplies for Vaccine Development
Product Literature
Customers Who Bought This Also Bought