Nalgene® Graduated Cylinders, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Graduated cylinders are suitable for food and beverage use.
- Excellent chemical resistance
- Made with permanent molded graduations (black on 1000mL and smaller sizes)
- No meniscus
- Blue polypropylene base is octagonal to prevent rolling and tipping
- Autoclaving may alter accuracy
Certifications: Cylinders meet accuracy requirements of ASTM Class B E1272 and ISO Standard 6706. Polypropylene resin meets requirements of CFR 21, Part 1787.1520 section of Food Additives Amendment of Federal Food and Drug Act.
Specifications
- Catalog No:
- 24776-188
- 24776-166
- 24776-100
- 24776-086
- 24776-144
- 24776-122
- 24776-020
- 24776-064
- 24776-042
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall