PYREX® Graduated Cylinders, Class A, To Contain, Serialized/Certified, with PYREX® Stopper, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, class A.
- Single metric scale cylinders are individually serialized
- Calibrated to contain (TC) in accordance with E-1272 and ASTM E-542
- White enamel graduations
PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Graduation markings are in durable white enamel. Single metric scale. With [ST] PYREX ®stopper.
Delivery: Supplied with a Corning Certificate of Identification and Capacity, traceable to NIST standards.
Specifications
- Catalog No:
- 24768-068
- 24768-046
- 24768-024
- 24768-150
- 24768-148
- 24768-126
- 24768-104
- 24768-090
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey