SP Wilmad-LabGlass Class B Graduated Cylinders with Standard Taper Stopper, TC, SP Industries
Supplier: Avantor
Certificates
About this item
These cylinders are calibrated "to contain" (TC) at 20 °C.
- Made of borosilicate glass
- Cylinders feature white graduations and a Standard Taper stopper
- Used to measure foam characteristics of emulsions and sedimentation values for solids
These laboratory, general-purpose grade graduated cylinders conform to ASTM E 1272 Style 2 Class B.
Specifications
- Catalog No:
- 14209-384
- Form:
- Short