Imhoff Triple Cone Support, Bennett Wood
Catalog #: 66190-009
Supplier: Bennett Wood Specialties
Specifications
- Description:Triple Cone Support
- Cat. no.:66190-009
- Supplier No.:6783
Specifications
About this item
Wood stand supports 3 Imhoff cones
Constructed so that cone graduation marks fall below the lower support deck for easy reading.
Customers Who Bought This Also Bought