Wood Funnel Support, Bennett Wood
Catalog #: 30580-001
Supplier: Bennett Wood Specialties
Specifications
- Description:Funnel Support
- Cat. no.:30580-001
- Supplier No.:2635
Specifications
About this item
Designed to hold six separatory funnels
With adjustable upper arm and auxiliary middle arm similarly slotted, and adjustable in height. Without glassware.
Customers Who Bought This Also Bought