Synthware Powder Funnels, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Funnels feature a 60° side angle and an inner [ST] joint.
- Ground glass joints
- Heavy wall borosilicate construction
- Various joint sizes available
These powder funnels are made of glass.
Customers Who Bought This Also Bought