Powder Funnels, Chemglass
Supplier: Chemglass
Certificates
About this item
Useful for the introduction of solids or liquids into reactions under atmospheric conditions
These powder funnels are made of glass.
Funnels have a 60° side angle and a [ST] inner joint.
Customers Who Bought This Also Bought