PYREX® Powder Funnels, Glass, Corning
Supplier: Corning
Certificates
About this item
PYREX®, borosilicate glass.
With short, wide stem. Used for transferring powders or filling bottles.
Specifications
- Catalog No:
- 89090-940
- 89091-142
- 89090-472
- 89090-626
- 89090-660
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Specifications
Related Information
Certificates
Search by Lot Number89090-472_09419056 89090-472_16117005 89090-626_28819013
Conformity declaration
Customers Who Bought This Also Bought