Synthware Powder Funnel with 60° Offset, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Funnels feature a 60° offset for use with multi-neck flasks.
- Ground glass joints
- Heavy wall borosilicate construction
- Various joint sizes available
- Offset Funnel
These powder funnel are made of glass.
Customers Who Bought This Also Bought