Synthware Powder Funnel with Flattened Side, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Funnel features a 60° angled side and a standard taper inner joint.
- Ground glass joints
- Heavy wall Borosilicate construction
- Various joint sizes available
- Flattened side wall
These powder funnel are made of glass.
Customers Who Bought This Also Bought