Goggle Conversion Kit, 506U
Catalog #: 77393-764
Supplier: UNIVET OPTICAL TECHNOLOGIES
New Product
Specifications
- Description:506U goggle conversion kit
Specifications
About this item
Kit is designed to convert 506 Value spectacles into a hybrid spectacle (goggle).
- Panoramic and wraparound lens for better side protection
- Adjustable elastic band
Soft rubber gasket provides gentle support on the face.
Customers Who Bought This Also Bought