5×6 X-Gen Over Spectacle
Catalog #: 77393-730
Supplier: UNIVET OPTICAL TECHNOLOGIES
New Product
Specifications
- Lens Coating:Anti-fog, scratch-resistant
- Lens Tint:Clear
- Frame Color:White/blue
Specifications
About this item
Light and comfortable safety eyewear, ideal for prolonged use.
- Anti-fog and anti-scratch lens treatment
- Temples with rubber terminals and tilting movement
- Adjustable temple length
- Soft and anatomic adjustable nose bridge
Designed to avoid annoying contact points with the face, these spectacles feature a frame with floating lens technology. VPlus anti-fog and anti-scratch enhancement is provided on the lenses.
Customers Who Bought This Also Bought