Face Shields, 711
Supplier: UNIVET OPTICAL TECHNOLOGIES
New Product
Certificates
About this item
The 711 series face shields feature an anti-fog lens treatment.
- Anti-fog lens treatment
- Disposable and interchangeable shields
- Ultra-lightweight design
- Multiple frame colors available
The 711 series face shields can be worn with prescription glasses and protective masks.
Ordering information: Anti-fog replacement shields are available separately.
Customers Who Bought This Also Bought