Individual Custom Prescription Insert for Univet 611 Goggles
Catalog #: 77393-708
Supplier: UNIVET OPTICAL TECHNOLOGIES
New Product
Specifications
- Description:Custom prescription insert
- Frame Color:Clear
Specifications
About this item
Custom Rx insert is designed for use with the univet 611 series goggle frame.
- Meets EN 166 standard
Custom prescription insert for 611 goggle shroud. The user must provide prescription details.
Customers Who Bought This Also Bought