Spare Anti-static Silicone Strap for 611/619 Series Goggles
Catalog #: 77393-706
Supplier: UNIVET OPTICAL TECHNOLOGIES
New Product
Specifications
- Description:Spare silicone strap
Specifications
About this item
Spare silicon strap is designed for use with the 611/619 series goggle frames.
- White strap
- Anti-static silicone construction
This spare silicone strap is designed for use with the Univet 611 and 619 series goggle shroud. Anti-static design with a buckle stopper built into the mold.
Customers Who Bought This Also Bought